Mobile Lab Island Instrument Carts, Lockable Cabinets, Polyethylene, TrippNT
Supplier: TrippNT
50591 50592 50593 51002
89085-584EA 4528.34 CAD
89085-584 89085-586 89085-588 10754-672 10754-674
Mobile Lab Island Instrument Carts, Lockable Cabinets, Polyethylene, TrippNT
Trolleys
Large bench, cabinet or instrument carts made from chemically resistant high density polyethylene for the demanding hospital and laboratory environment.
  • Large mobile benches easy to maneuver
  • 3 sizes
  • 2 colors
  • Easy open disappearing tambour drawer
  • Full extension pull out shelf
  • Bulk storage area
  • Chemically resistant polyethylene construction
  • Large flat work surface
  • 3 inch casters (2 locking)
  • Fully assembled, ready to use out of the box
  • In stock and ready to ship
  • American made by TrippNT
  • Total Replacement Lifetime Guarantee
These mobile benches or carts create extra flexible space in a work areas that are constantly changing.
The locking roll top (Tambour) door rolls up and out of the way to access shelves. The middle shelf pulls out on full extension drawer slides for easy access to supplies.