Mantle for Resin Reaction Flasks, Series O, Glas-Col®
Supplier: Glas-Col
100A O570 100AO572 100AO576 100AO578 100AO580
33794-045EA 1082.57 CAD
33794-045 33794-089 33794-169 33794-205 33794-249
Mantle for Resin Reaction Flasks, Series O, Glas-Col®
Heaters Heating Mantles
For resin reaction flasks from 500–4000mL capacity
Electrical: separable 1.2m (4') long, two-wire cord. For 115V operation. For 230V units, contact your Avantor representative.
Mantles provide even heat distribution and prevent scorching of clear resinous materials. Constructed of flexible fabrics for effective heating and protection of glass vessels. One-circuit heat input simplifies temperature control. Withstands 450°C (840°F) internal operating temperature.