Mantle for Round-Bottom Flasks, Series M, Glas-Col®
Supplier: Glas-Col
100CM96 100CM102 100CM106 100CM108 100CM110 100CM112 100CM114 100CM104 100CM116 100CM118
33778-027EA 1003.68 CAD
33778-027 33778-082 33778-129 33778-140 33778-162 33778-184 33778-209 33778-107 33778-220 33778-242
Mantle for Round-Bottom Flasks, Series M, Glas-Col®
Heaters Heating Mantles
Compact Series M mantles require little bench space
Mantle has a durable aluminum housing outside and resilient glass fabric inside to support flasks and protect flask walls from thermal strain. Only the bottom of the flask is covered so the contents being heated are fully visible. The mantle is grounded through the aluminum housing and requires a support. Internal operating temperature limit: 450°C (840°F). Electrical: 115V.
Ordering information: Supplied with a detachable 1.2m (4') long, three-wire cord and locking connector. For 230V units, contact your Avantor representative.