SHURSharp™ Microtome Blades, Triangle Biomedical
Supplier: General Data Healthcare
DMB-HD DMB-HP
CA27900-054CS 8724.11 CAD
CA27900-054 CA27900-058 CA27900-060
SHURSharp™ Microtome Blades, Triangle Biomedical
Microtomes and Microtome Assemblies Microtome Blades and Handles
SHURSharp™ offers three disposable microtome blades - a Teflon coated high-profile blade, a low-profile blade and heavy duty blade.
  • Proprietary coating process minimizes compression and produces excellent ribbon
  • SHURSharp™ low or high profile blades provide consistent sharpness and dependability
Blades are intended for use with the SHURSharp™ microtome blade holders. Low- and high-profile blades can also be used with most low/high-profile blade holders.
High-profile and low-profile blades will fit into any corresponding high or low profile bladeMore Product Information