Mantle/Controller Package, Series TM, Glas-Col®
Supplier: Glas-Col
100DTM106PL 100DTM112PL 100DTM102PL 100DTM104PL 100DTM108PL 100DTM110PL 100DTM114PL
33752-442EA 1670.69 CAD
33752-442 33752-500 33752-409 33752-420 33752-464 33752-486 33752-522
Mantle/Controller Package, Series TM, Glas-Col®
Heaters Heating Mantles
Glas-Col® offers TM series mantles and PowrTrol power control (see 33797-414 series) together in a special package
CSA certified.
The compact control is a solid-state, proportional voltage manual control that provides precise control of the output range, adjustable from 3–100%. Control has a lighted on/off power switch, three-wire load receptacle, and three-wire cord with molded plug. Series TM mantles feature an aluminum exterior with fabric interior, units up to 600W, 120V, 5A. Mantles withstand an internal operating temperature of 450°C (840°F).