Mantle for Spherical Flasks, Series TM, Glas-Col®
Supplier: Glas-Col
100BTM94 100BTM96 100BTM100 100BTM102 100BTM106 100BTM108 100BTM114 100BTM116 100BTM118 100BTM104 100BTM110 100BTM112
33752-059EA 754.01 CAD
33752-059 33752-061 33752-065 33752-067 33752-103 33752-125 33752-180 33752-205 33752-227 33752-089 33752-147 33752-169
Mantle for Spherical Flasks, Series TM, Glas-Col®
Heaters Heating Mantles
Spherical flask mantle covers only the flask’s bottom half, so contents are fully visible
Electrical: 115V operation (also available for 230V, except models 33752-059 and 33752-061). Supplied with detachable, 1.2m (4') long, three-wire cord and locking connector.
All units (except 33752-059, -061, -065) are CSA certified.
Durable aluminum exterior with fabric interior to softly nest glass vessels. Withstands 450°C (840°F) internal operating temperature. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full.