VWR® Atlas Pass-Through Overhead Storage Cabinets with Hinged Doors

Supplier: VWR International
VWR407530HG VWR407530HM VWR407531HM VWR407532HG VWR407532HM VWR407533HG VWR407533HM VWR407534HG VWR407534HM VWR407535HM VWR407536HG VWR407536HM
VWR407530HGEA 5823.75 CAD
VWR407530HG VWR407530HM VWR407531HM VWR407532HG VWR407532HM VWR407533HG VWR407533HM VWR407534HG VWR407534HM VWR407535HM VWR407536HG VWR407536HM
VWR® Atlas Pass-Through Overhead Storage Cabinets with Hinged Doors
Furniture Storage Cabinets
Steel cabinets are used in place of open shelving when enclosed storage and/or resistance to dust is required.

  • Steel fabrication
  • Hinged doors
  • Color: Light neutral

For use with VWR® Atlas mobile carts/mobile laboratory benches.

Ordering information: Supplied with two adjustable shelves and mounting hardware.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR