Nickel Chrome, gauze woven from wire 16 mesh
Supplier: JELLIFF CORP
16/020-4X4 16/020-5X5 16/020-6X6
66232-029PK 263.26 CAD
66232-029 66232-040 66232-062
Nickel Chrome, gauze woven from wire 16 mesh
Nickel Chrome
Non-rusting, high temperature
Withstands hotter flame than iron gauze. In squares of 16 mesh, No. 24 wire.