Regulator, Line, Diaphragm, Brass, Chrome Plated, Rotarex
Supplier: Ceodeux Inc.
SL1044 SL10116
89130-360EA 974.12 CAD
89130-360 89130-362
Regulator, Line, Diaphragm, Brass, Chrome Plated, Rotarex
Gas Regulators
Designed as a second stage line regulator for laboratory applications such as: gas delivery to inductive plasma spectrometer, protection and support gas for chromatograph, environmental emission monitoring, industrial hygiene or safety monitors and trace impurity analyzers.
As a second level of regulation,will supplya precise outlet pressure to the process. It can be used for many applications that need a high flow. Flexible wall or panel mounting possible with its compact design, the rear threads and fixing ring.
Pressure control with Balanced-Valve Technology: the effect of inletMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Body outlet 0.6 cm (1/4") NPT
Outlet type 0.6 cm (1/4") NPT
Seat material NBR Chrome-Plated Brass, FPM Stainless Steel