VWR® Cylinder Manifolds for High-Purity Noncorrosive and Corrosive Gases
Supplier: VWR International
82023-694 82023-702 82023-704 82023-708 82023-718 82023-722
82023-694EA 2417.42 CAD
82023-694 82023-702 82023-704 82023-708 82023-718 82023-722
VWR® Cylinder Manifolds for High-Purity Noncorrosive and Corrosive Gases
Gas Regulators
Manifolds are for use with cylinders with a maximum inlet pressure of 3000psig.
  • Ideal for CO2 Incubators
  • Brass or stainless steel parts depending on model
May be used with single- and two-stage general purpose, high-purity brass, high-purity stainless steel pressure regulators, as well as two-stage analytical pressure regulators. One-cylinder protocol stations include one 91.4 cm (36") pigtail; two-cylinder stations include two 91.4 cm (36") pigtails. Brass protocol stations feature brass wall brackets and stainless steel stations feature stainless steel wall brackets.
Ordering information: Regulators must be purchased separately.