Grade 226 Sheets, 580×580 mm

Supplier: Revvity Health Sciences
GR2266002
77661-494CS 553 CAD
77661-494
Grade 226 Sheets, 580×580 mm
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Revvity 226 filter paper sheets for component of medical device, blank device component.

  • Ahlstrom 226 filter paper is 100% cotton linter fiber filter paper
  • Blank, no printing

100% pure cotton linter filter paper provides even and uniform sample distribution. This feature delivers reliable and homogenous analytical results from across the dried sample area. Simply apply your sample, allow it to air dry completely, and then you are ready to ship or analyze.

Certifications: Components are not certified.

Packaging: Sold in packs of 100 sheets.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR