IVD-US Spot Saver Card

Supplier: Revvity Health Sciences
GRNK10022
77661-500CS 553 CAD
77661-500
IVD-US Spot Saver Card
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Revvity 226 blood collection device for general blood collection, excluding newborn screening - labeled for use in the USA.

  • Ahlstrom 226 filter paper is 100% cotton linter fiber filter paper designed to hold dried blood spot samples
  • Collection circles: 5 spots, 12.7 mm int.Ø no-tox ink
  • Flap to cover circles and tuck into card for safe transport
  • Barcode/serialization: Face and back for collecting/punching on either side, 8 digit code 39 barcode
  • Five-year expiration from date of production

100% pure cotton linter filter paper provides even and uniform sample distribution. This feature delivers reliable and homogenous analytical results from across the dried sample area. Being chemical free adds a level of safety and testing purity. Desiccation is not required before and/or after your sample has been applied in most situations. Simply apply your sample, allow it to air dry completely, and then you are ready to ship or analyze.

We can custom print and convert the grade 226 filter paper into a device form that meets your specialized requirements. Our capabilities also include printing barcodes, QR codes, and serialized numbers onto your devices to make them unique and traceable.

Certifications: FDA listed Class I Device.

Ordering information: If you are interested in newborn screening, please see GR2261006 and GR2261007 cards.

Packaging: Sold in packs of 100 cards.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR