Saddle Trax® 979 Anti Fatigue Mats

Supplier: NOTRAX USA, INC.
979R3675BL 979R3675GY 979R3675YB 979R4875BL 979R4875GY 979R4875YB 979R6075BL 979S0312GY
77661-466EA 6685.71 CAD
77661-466 77661-468 77661-470 77661-472 77661-474 77661-476 77661-478 77661-480
Saddle Trax® 979 Anti Fatigue Mats
Floor Mats Tacky Mats
Saddle Trax® is a thicker, heavier and stronger anti-fatigue floor mat. Engineered to meet the toughest requirements of today's industrial applications and is particularly suited for multi-shift operations in dry work areas where the worker is standing for prolonged periods of time; workstations, assembly lines.

  • Long-lasting vinyl top surface made from 100% recycled materials is combined with a dense closed cell PVC foam base
  • Diamond-plate surface texture provides added traction
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all four sides
  • Ergonomic, anti-fatigue and safety matting
  • Free of DMF, free of ozone depleting substances, free of silicone and free of heavy metals

At a full 1" thick, it is the ultimate anti-fatigue product maximizing worker productivity and comfort. The heavy-duty vinyl diamond-plate design provides non-directional traction, and is easy to clean. The top surface is combined with a dense closed cell foam base utilizing NoTrax® exclusive UniFusion™ technology. This virtually eliminates the possibility of de-lamination.

NoTrax® laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminates the slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete. The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat.

Environment: Dry areas
Surface: Industrial diamond plate
Backing: Closed cell polyvinylchloride with RedStop™ slip-resistant backing

Certifications: NSFI certified.

Caution: This product can expose you to chemicals including di-2-ethylhexyl phthalate and/or diisononyl phthalate which is known to the State of California to cause cancer, birth defects or reproductive harm.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR