VWR® M700 Portable pH/Ion/Conductivity/Dissolved Oxygen Meters
Supplier: SCHULER SCIENTIFIC
VWR-M700 77619-096 VWR-M700B 77619-100
77619-094EA 1644.96 CAD
77619-094 77619-096 77619-098 77619-100
VWR® M700 Portable pH/Ion/Conductivity/Dissolved Oxygen Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-M700 portable multi-parameter meter provides accurate and reliable measurements of pH, conductivity, ion, and dissolved oxygen within an easy to use meter.
  • High resolution LCD display (3.5")
  • Five-point calibration for pH with selectable pH buffer group
  • Built-in ion methods including F⁻, NH₄⁺ and NA⁺
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Three-point conductivity calibration with Standard Recognition
  • Dissolved oxygen calibration supports air-saturated water or zero oxygen
  • Ability to save 500 data sets for each parameter (GLP-compliant)
  • Rechargeable battery and Three-year meter warranty
The easy to use VWR-M700 portable meters provide a multitude of measurements with an easy to use meter. The large, backlit LCD display allows results to easily be viewed during operation. The carrying case allows for easy transport of the meter and accessories.
Ordering information: For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the Related Information section.
Delivery information: All meters include: carrying case, electrode holders (2), meter silicone rubber case, and hand strap.