Sartolab® P20 and P20 Plus Pressure Filtration Units
Supplier: Sartorius
18076----------N 18089----------D 18090----------D 18091----------D 18092----------D
CA77661-238PK 1260 CAD
CA77661-238 CA77661-240 CA77661-242 CA77661-244 CA77661-246
Sartolab® P20 and P20 Plus Pressure Filtration Units
Filters Membrane Filter Capsules
Sartolab® P20 and P20 Plus are ready-to-use pressure filter units for the clarification of 100 ml to 10 L particle-high-loaded solutions.
  • High flow rates
  • Large surface area (20 cm²)
  • Protein-preserving, low-binding membrane
  • Optimized membrane for low dead volume
  • Versions available with a prefilter for high particle load solutions
Sartolab® P20 and P20 Plus pressure filter units are available with a 0.2 µm or 0.45 µm polyethersulfone membrane, protected by a prefilter made of high purity quartz microfibers. Different configurations are available: sterile or non sterile, with or without PTFE automatic venting, with or without a filling bell (including cover) on the outlet, and with a combination of different inlet and outlet connectors to meet the needs of most applications.
Polyethersulfone (PES) is the membrane of choice for the Sartolab® P20 Plus pressure filter units, as it combines very low protein binding properties with the highest flow rates for filter batches from 100 ml to 10 L.