Front View Distillation Apparatus, Koehler
Supplier: Koehler Instrument
K45300 K45200 K45390 K45190 K45290 K45000 K45090
37000-276EA 23613.31 CAD
37000-276 37000-278 37000-280 37000-270 37000-282 37000-272 37000-284
Front View Distillation Apparatus, Koehler
Petroleum Testing Apparatus
The sample is evaporated and condensed under controlled conditions, with measurements of the temperatures at which various percentages are recovered and/or the percentages recovered at specified temperatures
Distillation apparatus are suitable for groups 0, 1, 2, and 3: motor and aviation gasolines, aviation turbine fuels, naphthas, kerosenes, distillate fuels, natural gasoline, liquid hydrocarbon mixtures, and other petroleum products. The 1000W heater unit and condenser are stainless steel.
Group 4 apparatus are additionally suitable for testing Grade No. 2 fuel oil, Grade No. 2-D diesel fuel oil, gas oils, and other distillates requiring condenser bath temperatures up to 60°C (140°F). Group 4 units feature a 300W copper immersion condenser heater.
Units feature fully insulated condenser and heater units with stepless electronic control, flask support platform, viewing window, and rack and pinion heater-elevation mechanism with push-turn control knob.
Certifications: Meet specifications: ASTM D86, D216, D233, D447, D850, D1078, E133; IP 123, 195; ISO 3405; DIN 51751; FTM 791-1001, 791-1015; NF M 07-002.
Ordering information: Supplied with graduate support block and flask support boards.