Genomic DNA Blood/Cultured Cell Kits, IBI Scientific
Supplier: IBI Scientific
Kits provide a fast and economical method for the purification of total DNA (including genomic, mitochondrial, and viral DNA) fresh whole blood, plasma, serum, buffy coat, other bodily fluids, lymphocytes, bacteria, and cultured cells
The method uses a chaotropic salt to lyse cells and degrade proteins. The DNA in the chaotropic salt is bound by the glass fiber matrix of the spin column. Once any contaminates have been removed, the purified DNA is eluted by a low salt elution buffer or water. The entire procedure can be completed in 40 minutes (MINI) or 60 minutes (MAXI) without the use of phenol extraction or alcohol precipitation.
Kit sample size: Up to 300 µL of fresh whole blood; up to 107 animal cultured cells
Caution: For research use only. Components contain irritant agents. Always wear a lab coat, disposable gloves, and protective goggles during use.