Bench-Mounted Secondary Vacuum Trap, Labconco®
Supplier: Labconco
7538000 7538200 7538400
26673-353EA 2970.4 CAD
26673-353 26673-355 26673-357
Bench-Mounted Secondary Vacuum Trap, Labconco®
Freeze Dryers
Provides efficient processing of samples with low eutectic points
The secondary trap with dry ice and solvent in its well cools to approximately –75°C (–103°F). The vacuum trap used alone serves as an economical condenser. Simply connect a sample directly to the incoming port and add a vacuum pump to the outgoing port. Inner pot holds either 1.8 or 2.85L of dry ice and alcohol.