Resolve™ Immersion Oil, Epredia™
Supplier: RICHARD-ALLAN SCIENTIFIC CA
M2000 M2004 M2000 M3000 M5000 M2004 M3004 M4004 M5004 M4000
48218-049CS 290.05 CAD
48218-049 48218-402 CA48218-049 CA48218-061 CA48218-106 CA48218-402 CA48218-424 CA48218-445 CA48218-467 CA-48218-081
Resolve™ Immersion Oil, Epredia™
Stains Microscopy and staining reagents
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ immersion oil, which contains no PCBs.
  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes
  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150
It is a clear, colorless oil that allows light transmission throughout most of the UV-VIS spectrum.