Ramsbottom Carbon Residue Apparatus, Koehler
Supplier: Koehler
K27100 K27190
37000-182EA 24048.52 CAD
37000-182 37000-184
Ramsbottom Carbon Residue Apparatus, Koehler
Petroleum Testing Apparatus
Provides an indication of the deposit-forming tendencies of fuels by determining the volume of carbon residue left after evaporation and pyrolysis of a sample oil
Cast-iron block type coking furnace features microprocessor temperature control with °C/°F switchable digital set point and display. Stainless steel cabinet is heavily insulated with a three-layer refractory top for excellent heat retention. Built-in control circuit automatically interrupts power if the block temperature exceeds the programmed cutoff point.
Certifications: Conforms to specifications: ASTM D524, D6074; IP 14;More Product Information