Compact Glove Boxes, Plas-Labs™
Supplier: Plas-Labs
830-ABB/EXP 830-ABD/EXP 830-ABC/EXP 830CP-10/EXP 830CP-11/EXP 830CP-20/EXP 830CP-21/EXP 830CP-30/EXP 830CP-31/EXP
CA10718-112EA 69175.83 CAD
CA10718-112 CA10718-114 CA32940-008 CA97000-994 CA97000-996 CA97000-998 CA97001-000 CA97001-002 CA97001-004
Compact Glove Boxes, Plas-Labs™
Glove Boxes and Glove Bags
Designed specifically for use when weighing hazardous materials that must be contained, glove boxes provide cleanliness, safety, portability, and a draft-free atmosphere for the maximum effectiveness of your balance
Top section is removable for easy installation of equipment, such as analytical balances, top loaders, bench scales, and moisture analyzers. Basic features include four purging gas valves (two on the transfer chamber and two on the main chamber), a white leveling tray for easy transferring of liquids, white ambidextrous Hypalon® gloves for superior chemical and UV resistance,More Product Information