Cloud and Pour Point Apparatus, Koehler
Supplier: Koehler
K46100 K46195 K46196 K46300 K46395 K46396 K46001
37000-292EA 46740.08 CAD
37000-292 37000-294 37000-296 37000-298 37000-300 37000-302 37000-290
Cloud and Pour Point Apparatus, Koehler
Petroleum Testing Apparatus
Devices indicate the lowest temperature of utility for petroleum products
Mechanically refrigerated units feature multiple four-jacket baths, each preset at a different temperature for convenient cloud and pour point testing. Bench models have three baths, set at –1, –18, and –35°C (30, 0, and –30°F). Floor models have four baths, set at –1, –18, –35, and –51°C (30, 0, –30, and –60°F). Each bath has a phenolic top plate with ports for a thermometer and four test jackets. Synthetic sponge top plate covers and gasketed bath hoods deter ice accumulation. Bath cabinet and interior are constructed of stainless steel. Floor models ride on swivel casters.
The cloud and pour point chamber immerses four copper test jackets in suitable freezing mixtures, providing inlet and outlet connections for circulating refrigerated coolant from an external source. Consists of steel exterior housing with polyurethane enamel finish and all-copper interior for corrosion resistance. Removable top plate and 13mm (1/2") cork insulation aid in cold retention.
Certifications: Conform to specifications: ASTM D97, D2500, D6074, D6158; IP 15, 219; ISO 3015, 3016; DIN 51597; FTM 791-201; NF T 60-105.
Ordering information: Chamber and baths are supplied with test jackets, gaskets, disks, and thermometer holders for test jars and cooling baths. Test jars and thermometers are not included. A software package is also available for unit 37000-292; contact your Avantor representative for more information.