VWR® 12-Channel Polypropylene Reagent Reservoirs
Supplier: VWR International
77575-240 77575-244 77575-254 77575-258
77575-240CS 76.26 CAD
77575-240 77575-244 77575-254 77575-258
VWR® 12-Channel Polypropylene Reagent Reservoirs
Reagent Reservoirs
These 12-channel reservoirs are made from high-quality, chemical resistant PP. Designed for serial dilutions or pipetting individual reagents using up to 12-channel pipetters.
  • V-shaped basins facilitate easy withdrawal of sample solutions
  • Each channel holds up to 3 ml of solution and is individually numbered
  • Includes a tight-fitting lid
  • Individually wrapped packaging
  • Non-sterile or sterilized by irradiation, SAL 10⁻⁶ (ISO 11137)
  • Lot-certified, non-pyrogenic, DNase- and RNase-free
12-channel reagent reservoirs feature twelve 3 ml compartments, designed with V-shaped channels to maximize complete sample withdrawal. They are ideal for all liquid handling applications.
Certifications: ISO 13485 and ISO 9001.