Nitrogen Dry Glove Boxes, Plas-Labs™
Supplier: Plas-Labs
850-NB/EXP 850-NBB/EXP
CA87000-724EA 109772.3 CAD
CA87000-724 CA87000-726
Nitrogen Dry Glove Boxes, Plas-Labs™
Glove Boxes and Glove Bags
Glove boxes provide completely enclosed chambers ideal for working in a controlled atmosphere
Glove boxes have a one-piece clear acrylic top and die-molded thermoset white bottom. Units feature two vacuum pressure pumps, one each for the transfer chamber and drying train; the transfer chamber has an adjustable vacuum gauge. The drying train includes polycarbonate canisters containing DesiSphere® desiccant/purifier. Self-sealing quick disconnects enable the operator to change the drying train without disturbing the internal atmosphere. Each glove port is fitted with an ambidextrous glove made of Hypalon™ material for superior chemical and UV resistance. Four ground key cock valves allow purging. All clamps are adjustable to compensate for wear. Front viewing panel is sloped to minimize glare. Units have a bright fluorescent lighting system and illuminated controls.
Units are UL listed, CSA certified, and CE marked.
Inert gases such as argon, nitrogen, or plasma are easily maintained.
Ordering information: Supplied with a one-year manufacturer’s warranty on parts. Units with a 220V configuration are available; contact your Avantor sales representative for more information.