VWR® Wire Shelving with Round Posts
Supplier: VWR International
89409-900 89409-902 89409-904 89409-906 89409-908 89409-910 89409-912 89409-914 89409-916 89409-918 89409-920 89409-922 89409-924 89409-926 89409-928 89409-930 89409-932 89409-934 89409-936 89409-938 89409-940 89409-942 89409-944 89409-946 89409-948 89409-950 89409-952 89409-954 89409-956 89409-958 89409-960 89409-962 89409-964 89409-966 89409-968 89409-970 89409-972 89409-974 89409-976 89409-978 89409-980 89409-982 89409-984 89409-986 89409-988 89409-990 89409-992 89409-994 89409-996 89409-998 89410-000 89410-002 89410-004 89410-006 89410-008 89410-010 89410-012 89410-014 89410-016 89410-018 89410-020 89410-022 89410-023 89410-024 CA89409-900 CA89409-902 CA89409-904 CA89409-906 CA89409-908 CA89409-910 CA89409-912 CA89409-914 CA89409-916 CA89409-918 CA89409-920 CA89409-922 CA89409-924 CA89409-926 CA89409-928 CA89409-930 CA89409-932 CA89409-934 CA89409-936 CA89409-938 CA89409-940 CA89409-942 CA89409-944 CA89409-946 CA89409-948 CA89409-950 CA89409-952 CA89409-954 CA89409-956 CA89409-958 CA89409-960 CA89409-962 CA89409-964 CA89409-966 CA89409-968 CA89409-970 CA89409-972 CA89409-974 CA89409-976 CA89409-978 CA89409-980 CA89409-982 CA89409-984 CA89409-986 CA89409-988 CA89409-990 CA89409-992 CA89409-994 CA89409-996 CA89409-998 CA89410-000 CA89410-002 CA89410-004 CA89410-006 CA89410-008 CA89410-010 CA89410-012 CA89410-014 CA89410-016 CA89410-018 CA89410-020 CA89410-022 CA89410-023 CA89410-024
89409-900EA 477.29 CAD
89409-900 89409-902 89409-904 89409-906 89409-908 89409-910 89409-912 89409-914 89409-916 89409-918 89409-920 89409-922 89409-924 89409-926 89409-928 89409-930 89409-932 89409-934 89409-936 89409-938 89409-940 89409-942 89409-944 89409-946 89409-948 89409-950 89409-952 89409-954 89409-956 89409-958 89409-960 89409-962 89409-964 89409-966 89409-968 89409-970 89409-972 89409-974 89409-976 89409-978 89409-980 89409-982 89409-984 89409-986 89409-988 89409-990 89409-992 89409-994 89409-996 89409-998 89410-000 89410-002 89410-004 89410-006 89410-008 89410-010 89410-012 89410-014 89410-016 89410-018 89410-020 89410-022 89410-023 89410-024 CA89409-900 CA89409-902 CA89409-904 CA89409-906 CA89409-908 CA89409-910 CA89409-912 CA89409-914 CA89409-916 CA89409-918 CA89409-920 CA89409-922 CA89409-924 CA89409-926 CA89409-928 CA89409-930 CA89409-932 CA89409-934 CA89409-936 CA89409-938 CA89409-940 CA89409-942 CA89409-944 CA89409-946 CA89409-948 CA89409-950 CA89409-952 CA89409-954 CA89409-956 CA89409-958 CA89409-960 CA89409-962 CA89409-964 CA89409-966 CA89409-968 CA89409-970 CA89409-972 CA89409-974 CA89409-976 CA89409-978 CA89409-980 CA89409-982 CA89409-984 CA89409-986 CA89409-988 CA89409-990 CA89409-992 CA89409-994 CA89409-996 CA89409-998 CA89410-000 CA89410-002 CA89410-004 CA89410-006 CA89410-008 CA89410-010 CA89410-012 CA89410-014 CA89410-016 CA89410-018 CA89410-020 CA89410-022 CA89410-023 CA89410-024
VWR® Wire Shelving with Round Posts
Furniture Shelves
Starter units include four round corner posts and four shelves, and add-on units include two posts, four shelves and clips.
  • Starter units include four round corner posts and four shelves
  • Chrome finish
  • Truss-reinforced edges
Chrome finish provides improved corrosion resistance. Each shelf features truss-reinforced edges for exceptional durability and can support up to 800 lbs. when weight is evenly distributed. Posts feature groove rings in 1 increments for shelf height adjustability.
Ordering information: Shipped unassembled for compact storage.