Protector® Glove Box Base Stands, Labconco®
Supplier: Labconco
Protector®
5062020
15203-116EA 11281.2 CAD
15203-116
Protector® Glove Box Base Stands, Labconco®
Glove Boxes and Glove Bags
Epoxy-coated, welded steel base stands have a permanently attached, black phenolic laminate work surface
Stands for single glove boxes have a work surface measuring 152.4Wx78.7Dx2.8Hcm (60x31x11/8"). Height is adjustable from 83.8–101.6cm (33–40") high. Mobile stand 97000-032 features 12.7cm (5") swivel casters and locking brakes. Fixed stand 97000-034 has adjustable leveling feet.
Stand 15203-116 supports a double glove box. Work surface measures 264.1Wx78.7Dx2.8Hcm (104x31x11/8"). Height is adjustable from 83.8–101.6cm (33–40") high with swivel casters.
Lower shelf provides convenient space for an accessory vacuum pump or optional drying train.