VWR® Reagent Reservoirs
Supplier: VWR International
77575-238 77575-242 77575-246 77575-248 77575-250 77575-252 77575-256 77575-278 77575-280 77575-282 77575-284 77575-286 77575-260 77575-262 77575-264 77575-266 77575-268 77575-270 77575-272 77575-274 77575-276
77575-238CS 176.49 CAD
77575-238 77575-242 77575-246 77575-248 77575-250 77575-252 77575-256 77575-278 77575-280 77575-282 77575-284 77575-286 77575-260 77575-262 77575-264 77575-266 77575-268 77575-270 77575-272 77575-274 77575-276
VWR® Reagent Reservoirs
Reagent Reservoirs
Reagent reservoirs are designed for efficient liquid transfer, making them an ideal choice for repetitive pipetting tasks. These reagent reservoirs for multichannel pipettes provide a trough wide enough to fit 8 and 12 channel pipettes.
  • Trough design with graduations on the inside wall and pour-off spouts on four corners
  • Slightly tilted inner surface helps reduce residue
  • Non sterile or sterilized options available, achieving SAL 10⁻⁶ (ISO 11137)
  • Reservoirs are lot-certified, non-pyrogenic, DNase- and RNase-free
VWR® reagent reservoirs are an excellent choice for holding different reagents, including those used in the setup of high-throughput screening, ELISA, PCR, and cell culture experiments.
The 25 ml reservoirs are available with a single well or 3-well design (each holding 5 ml). The 50 ml reservoirs are available in PVC, PET, and PS.
Certifications: ISO 13485 and ISO 9001.
Packaging: Available with inner packaging formats of 1/bag, 5/bag, and bulk.