Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tubes
Supplier: Thermo Fisher Scientific
3400-DLB 3401-DLB 3402-DLB 3403-DLB 3404-DLB 3405-DLB
77675-224CS 749.55 CAD
77675-224 77675-226 77675-228 77675-230 77675-232 77675-234
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
These tubes are low DNA binding, ensuring minimal sample loss, and free from PCR inhibitors, UV leachables, and human DNA, making them ideal for sensitive applications. Furthermore, the Sustain Series are sustainable – with biobased resin and ISCC certification, underscoring their eco-friendly design. The standard DNA low binding tubes ensure minimal sample loss, and free from PCR inhibitors, UV leachables, and human DNA, making them ideal for sensitive applications.
  • Low DNA binding
  • PCR inhibitor free
  • UV leachables free
  • Human DNA free
  • Sustain series are sustainable with biobased resin and ISCC certification
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results. Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.