EasySep™ Direct HLA Crossmatch B Cell Isolation Kits
Supplier: STEMCELL Technologies
19684 19684RF
MSPP-19684EA 1164.62 CAD
MSPP-19684 MSPP-19684RF
EasySep™ Direct HLA Crossmatch B Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human B cells for HLA analysis
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 99.8% purity of isolated cells
  • Fast, easy-to-use, and column-free
  • Isolated cells are untouched
The EasySep™ Direct HLA Crossmatch B Cell Isolation Kit can be used to isolate highly purified B cells directly from human whole blood, spleen, or lymph nodes by immunomagnetic negative selection. This cell isolation kit targets non-B cells for removal with antibodies recognizing specific surface markers. Unwanted cells are labeled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry crossmatch.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').