HyFlex® 11-561 Cut-Resistant Gloves, Vend Pack
Supplier: Ansell Canada
11561VP050 11561VP060 11561VP070 11561VP080 11561VP090 11561VP100 11561VP110 11561VP120
CA76519-332CS 3318.35 CAD
CA76519-332 CA76519-334 CA76519-336 CA76519-338 CA76519-340 CA76519-342 CA76519-344 CA76519-346
HyFlex® 11-561 Cut-Resistant Gloves, Vend Pack
Gloves
Ansell's lightest EN ISO cut C and ANSI/ISEA 105-2024 cut A3 glove (versus standard EN ISO cut C and ANSI/ISEA 105-2024 cut A3-rated gloves) offering two times greater cut resistance versus similar gloves made of standard HPPE yarn.
  • Ultra-lightweight design
  • 20% lighter for unparalleled comfort and dexterity
  • 2x greater cut resistance
  • Improved FORTIX™ technology offering up to 20% greater durability
The ultra-lightweight design of the HyFlex® 11-561 makes this EN ISO Cut C and ANSI/ISEA 105-2024 A3-rated safety glove 20% lighter. The gloves provide heightened defense, with two times greater cut resistance using next generation single HPPE yarn developed with INTERCEPT™ cut resistance technology for reliable hand protection. Offering increased durability for extended use, improved FORTIX™ technology delivers up to 20% more abrasion resistance for longer lasting handling in abrasive conditions.
HyFlex 11-561's palm-dipped coating improves grip, while a reinforced thumb crotch defends against rapid wear. Certified harm-free, Dermatest® and Oeko-Tex® certification ensures these PPE gloves are skin-friendly and free of harmful substances. Touchscreen compatibility allows the wearer to work with multiple touchscreen devices while keeping gloves on.
Certifications: ANSI/ISEA 105-2024 cut level A3, ANSI/ISEA 105-2024 ABR level 4, ANSI/ISEA 105-2024 PUN level 4, dermatologically tested, ecovadis label, EN 388:2016, EN ISO 21420:2020, food contact, OEKO-TEX® centexbel, category II.
Packaging: Gloves are packaged six pairs per bag, 144 pairs per carton.