RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktails
Supplier: STEMCELL Technologies
15026 15066
MSPP-15026-40EA 536.92 CAD
MSPP-15026-40 MSPP-15066
RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktails
Cell Separation Media
Immunodensity isolation of untouched stem/progenitor cells from cord blood.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail is designed to isolate progenitor cells from cord blood and whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD61, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (STMC07801), the unwanted cells pellet along with the RBCs. The purified progenitor cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium. If using large volumes of blood, we recommend the RosetteSep™ Cord Blood Progenitor Enrichment Kit with HetaSep™ (STMC15276)
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).