KT™ KT5+ Premium Advanced Adhesive Slides
Supplier: StatLab
KT5+-GY-90 KT5+-GY-CL KT5+-LV-90 KT5+-LV-CL KT5+-OR-90 KT5+-OR-CL KT5+-PK-90 KT5+-PK-CL KT5+-TN-90 KT5+-TN-CL KT5+-WH-90 KT5+-WH-CL KT5+-YL-90 KT5+-YL-CL KT5+-BL-90 KT5+-BL-CL KT5+-GR-90 KT5+-GR-CL
77661-608CS 1303.56 CAD
77661-608 77661-610 77661-612 77661-614 77661-588 77661-590 77661-592 77661-594 77661-596 77661-598 77661-600 77661-602 77661-604 77661-606 77661-580 77661-582 77661-584 77661-586 77661-589 77661-591 77661-593 77661-595 77661-597 77661-599 77661-601 77661-603 77661-605 77661-607 77661-609 77661-611 77661-613 77661-615 77661-581 77661-583 77661-585 77661-587
KT™ KT5+ Premium Advanced Adhesive Slides
Slides Microscope Slides
KT™ Slides KT5+ Premium German advanced adhesion slides are an excellent choice for H and E, special stains, frozen sections, and even the toughest IHC procedures with difficult tissues.
  • PiSmart S2 slide printer validated and compatible with both thermal transfer and UV laser printers
  • Colored marking areas for efficient specimen color-coding, identification, labeling, and printing
  • Superior adhesion maintains tissue integrity throughout the toughest of tissues or protocols
  • No background staining
  • Hybrid waterbath behavior: the tissue section jumps to the slide but water remains underneath for repositioning if necessary
Available in nine unique colors, in both clipped and 90° corners. Cold room storage is recommended.
Packaging: Sold by the case. 144 slides per gross, 10 gross/case.