KT™ KT3+ Premium Standard Adhesive Slides
Supplier: StatLab
KT3+-GR-90 KT3+-GR-CL KT3+-GY-90 KT3+-GY-CL KT3+-LV-90 KT3+-LV-CL KT3+-OR-90 KT3+-OR-CL KT3+-PK-90 KT3+-PK-CL KT3+-TN-90 KT3+-TN-CL KT3+-WH-90 KT3+-WH-CL KT3+-YL-90 KT3+-YL-CL KT3+-BL-90 KT3+-BL-CL
77661-548CS 988.51 CAD
77661-548 77661-550 77661-552 77661-554 77661-556 77661-558 77661-560 77661-562 77661-564 77661-566 77661-544 77661-546 77661-568 77661-570 77661-572 77661-574 77661-576 77661-578 77661-549 77661-551 77661-553 77661-555 77661-557 77661-559 77661-561 77661-563 77661-565 77661-567 77661-569 77661-571 77661-573 77661-575 77661-577 77661-579 77661-545 77661-547
KT™ KT3+ Premium Standard Adhesive Slides
Slides Microscope Slides
KT™ Slides KT3+ Premium German Standard Adhesion Slides are an excellent choice for H and E, special stains, frozen sections, and IHC procedures.
  • PiSmart S2 slide printer validated and compatible with both thermal transfer and UV laser printers
  • Colored marking areas for efficient specimen color-coding, identification, labeling, and printing
  • Excellent adhesion, maintaining tissue integrity through tough chemical processing
  • Hydrophobic waterbath behavior: tissue sections easily jump on the slide
Available in nine unique colors, in both clipped and 90° corners. Cold room storage is recommended.
Packaging: Sold by the case. 144 slides per gross, 10 gross/case.