Arium® Mini Plus Extend Lab Water Purification System

Supplier: Sartorius
H2O-MAR-T H2O-MAR-UV-T
CA77627-812EA 8968.75 CAD
CA77627-812 CA77627-814
Arium® Mini Plus Extend Lab Water Purification System
Water Purification Systems Pure and Ultra Pure Water Systems
The Arium® Mini Plus Extend is a water purification system from potable tap water to Type I water. It combines maximum flexibility with a compact design, featuring a removable dispensing unit for convenience without compromising on consistently high water quality for up to 10 L per day.

  • Easily removable dispensing unit with magnetic mount for ergonomic and splash-free filling into a wide variety of containers
  • Manual and volume-controlled water dispensing
  • Dispensing of the last dispensed volume
  • Can be combined with sterile plus final filter or CellPlus filter
  • Installation option: Installation option

The well-known Sartorius bag tank technology with a five-liter disposable bag is used, requiring no cleaning. Available with or without an integrated UV lamp, depending on your specific application needs.

Popular applications for Arium® mini plus extend
-Preparation of samples, media and buffers
-Cell and tissue cultures
-Gas chromatography

Application:
-GC-MS; ELISA
-Preparation of reagents
-IC; HPLC/UHPLC
-Buffer and media prep
-AAS
-Histology; Photometry
-TOC-analysis
-Feed lab water systems
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR