Pierce MagnaBind™ Beads and Supports, Thermo Scientific
Supplier: Thermo Scientific
21344 21348 21349 21352 21353 21354 21356
CAPI21344EA 1391.26 CAD
CAPI21344 CAPI21348 CAPI21349 CAPI21352 CAPI21353 CAPI21354 CAPI21356
Pierce MagnaBind™ Beads and Supports, Thermo Scientific
Beads Magnetic Beads
Magnetic beads are a convenient affinity support for a variety of assays, which allow easy purification of the target without columns or centrifugation
  • Available pre-coated with popular affinity ligands or derivatized for covalent attachment of proteins and other specific ligands
  • Beads do not irreversibly aggregate because they have no magnetic memory; collect and disperse the beads multiple times, if needed
  • Most separations require a short 5 to 10 minute bench-top procedure
MagnaBind™ beads are available pre-coated with Protein A, Protein G, streptavidin, anti-mouse or anti-rabbit antibodies. Activated beads, with either amine or carboxyl groups, are also available for attaching other proteins or affinity ligands to a magnetic particle.
After a binding step in an affinity purification procedure, the magnetic particles are easily and rapidly collected by placing the microcentrifuge tube or reaction vessel next an appropriate rare-earth magnet. MagnaBind™ beads respond rapidly to MagnaBind™ magnets but can be easily dispersed and regathered multiple times (ie. they will not irreversibly aggregate) because they do not have any magnetic memory.
Particles: 1 x 10⁸/mg
Surface area: >100 m²/g
Ø: 1-4 µm