VWR® Generator Back-Up Systems
Supplier: VWR International
89080-023 89080-025 89080-027 89080-029 89080-031 89080-033 89080-035 89080-037 89080-039 89080-041 89080-043 89080-045
89080-023EA 3084.21 CAD
89080-023 89080-025 89080-027 89080-029 89080-031 89080-033 89080-035 89080-037 89080-039 89080-041 89080-043 89080-045
VWR® Generator Back-Up Systems
Gas Cylinders
Model SG 770 generator back-up systems provide continuous backup supply of gas in case of generator failure or power outage.
  • Continuous backup supply of gas
  • Features diaphragm isolation valves with high leak integrity
  • Adjustable gas service
Cylinder turns on automatically when generator pressure drops below preset value, and turns off when generator gas supply increases to standard level. Features 0–125psig line regulator, diaphragm isolation valves with high leak integrity, 91.4 cm (36") flexible pigtail hose with integrated reverse flow check valve, 1/4-turn isolation shut off valves, 0.6 cm (1/4") Swagelok® inlet (with reverse flow check valve) and outlet connections, adjustable gas service, and PTFE seals and seat.