VWR® Calibrated Dry Block/Incubator Liquid-In-Glass Thermometers
Supplier: VWR International
89082-190 89082-192
89082-190EA 496.03 CAD
89082-190 89082-192
VWR® Calibrated Dry Block/Incubator Liquid-In-Glass Thermometers
Thermometers Analog Thermometers
Individually calibrated thermometers feature blue organic liquid fill against lead-free white back glass.
  • Ideal for use in all standard heating blocks and water baths
  • Individually serialized
Each thermometer is individually calibrated against equipment whose calibration is traceable to the National Institute of Standards and Technology.
Ordering information: Includes an individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.