VWR® Zero Air Generators
Supplier: VWR International
CA26000-024 26000-024 26000-026 26000-028 89237-566
CA26000-024EA 21378.46 CAD
CA26000-024 26000-024 26000-026 26000-028 89237-566
VWR® Zero Air Generators
Gas Generators Zero Air Generators
Systems produce UHP zero air with a purity level below 0.05ppm total hydrocarbon content from a compressed air supply.
  • Eliminate dangerous gas cylinders from lab
  • Recommended and used by major GC and column manufacturers
  • Generated air can be used as a support gas
  • Simple installation with standard equipment
The air generated can be used as support gas for total hydrocarbon analyzers, as nebulizer and exhaust pump gas for LC/MS instruments, or as oxidants/support gas for GC with FID, FPD, and NPD detectors. The generators are engineered for easy installation, operation, and long-term performance. All that is required for installation is a standard compressed air line and an electrical outlet.