VWR® Membrane Nitrogen Generators
Supplier: VWR International
CA26000-014 CA26000-016 CA26000-018 26000-014 26000-016 26000-018
CA26000-014EA 11947.23 CAD
CA26000-014 CA26000-016 CA26000-018 26000-014 26000-016 26000-018
VWR® Membrane Nitrogen Generators
Gas Generators Nitrogen Generators
VWR® Membrane Nitrogen Generators are designed to supply single or multiple LC/MS instruments with dry nitrogen at purities of 99.0–99.5%.
  • Recommended and certified by all major LC/MS instrument manufacturers
These membrane generators can also be used for solvent evaporation or analytical instruments. Installation requires a minimum of 60psig of compressed air to a 63.5mm (1/4") or 127mm (1/2") inlet connection. The outlet nitrogen supply is then directed to analytical instruments. The generators require no electrical connections and the onlyMore Product Information