Dessicant Packet, Qiagen
Supplier: Qiagen
WB100024 WB100024 WB100003
14222-840EA 835.03 CAD
14222-840 CA14222-838 14222-838
Dessicant Packet, Qiagen
Desiccators Desiccants
  • Ensure that FTA Cards remain dry during transport or storage
  • Change from orange to green to indicate absorption of moisture