Fast-Ion Maxi Plasmid Kits, IBI Scientific

Supplier: IBI Scientific

IB47120 IB47121 IB47122
95039-660EA 173.63 CAD
95039-660 95039-662 95039-664
Fast-Ion Maxi Plasmid Kits, IBI Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

IBI’s Maxi Plasmid Kits use pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures


  • Sample Size: 100ml high-copy plasmid/250ml low-copy plasmid
  • Operation: Gravity Flow Column
  • Expectant Yield: 500µg to 1mg of plasmid DNA
  • Operation Time: 120 minutes or less


The Maxi Plasmid Kits use pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures. In this process, the modified alkaline lysis method and RNase treatment are used for creating cleared lysate with minimal genomic DNA and RNA contaminants. Using a gravity-flow procedure, the plasmid DNA in crude lysate has been bound to the column. The contaminants can be washed off with a wash buffer. Finally, the purified DNA is eluted by a high salt buffer and then precipitated with isopropanol for desalting. The entire procedure can be completed in less than two hours. The purified plasmid DNA is suitable for transfection, sequencing reactions, PCR, and in-vitro transcription.

Order Now

SPECIFICATIONS

Description Maxi Fast-Ion Plasmid Sample Kit Maxi Fast-Ion Plasmid Kit
Environmentally Preferable
Application
Target molecule Plasmid Plasmid
Refine target molecule Plasmid,Larger vectors (Cosmid, BAC, PAC, Fosmid) Plasmid,Larger vectors (Cosmid, BAC, PAC, Fosmid)
Sample type Bacteria Bacteria
Method / Format Column (anion exchange) Column (anion exchange)
Prep size Maxi Maxi
Yield 500 µg to 1 mg of Plasmid DNA 500 µg to 1 mg of Plasmid DNA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR