E.Z.N.A.® MicroElute® Cleanup Kits, Omega Bio-tek
Supplier: OMEGA BIO-TEK CA
R6247-00 R6247-01
CA101319-098EA 35.31 CAD
CA101319-098 CA101319-100 CA101319-102 CA101319-188 CA101319-190
E.Z.N.A.® MicroElute® Cleanup Kits, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Achieve fast and reliable purification of DNA or RNA with a relatively small elution volume of 10–15µL
Kits do not use organic extractions or alcohol precipitations to allow for safe reactions that can be completed in less than 15 minutes. In addition, the kits yield 80–90% recovery of DNA or RNA fragments. To recover DNA from enzymatic reactions, use the DNA cleanup kits. To recover DNA from PCR reactions, use the Cycle-Pure kits. To recover DNA from agarose, use the gel extraction kits. The RNA kits use a high salt buffer system to bind RNA molecules greater than 200 bases to a matrix, while removing other contaminants.