VWR® Atlas Shelf Brackets
Supplier: VWR International
VWRBH12 VWRBH15 VWRBH18 VWRBH24 VWRBHC12 VWRBHC15 VWRBHC18 VWRBHC24 VWRBL12 VWRBL15 VWRBL18 VWRBL24 VWRBLC12 VWRBLC15 VWRBLC18 VWRBLC24 VWRBLC36 VWRBU12 VWRBU18 VWRBU24 VWRBU36 VWRBH12-W VWRBHC12-W VWRBL12-W VWRBU24-W VWRBLC15-W VWRBU36-W VWRBHC18-W VWRBL24-W VWRBU18-W VWRBU12-W VWRBH18-W VWRBH24-W VWRBLC24-W VWRBHC15-W VWRBL15-W VWRBH15-W VWRBLC18-W VWRBLC12-W
VWRBH12PK 263.37 CAD
VWRBH12 VWRBH15 VWRBH18 VWRBH24 VWRBHC12 VWRBHC15 VWRBHC18 VWRBHC24 VWRBL12 VWRBL15 VWRBL18 VWRBL24 VWRBLC12 VWRBLC15 VWRBLC18 VWRBLC24 VWRBLC36 VWRBU12 VWRBU18 VWRBU24 VWRBU36 VWRBH12-W VWRBHC12-W VWRBL12-W VWRBU24-W VWRBLC15-W VWRBU36-W VWRBHC18-W VWRBL24-W VWRBU18-W VWRBU12-W VWRBH18-W VWRBH24-W VWRBLC24-W VWRBHC15-W VWRBL15-W VWRBH15-W VWRBLC18-W VWRBLC12-W
VWR® Atlas Shelf Brackets
Furniture Shelves
Designed to be infinitely adjustable along vertical Atlas uprights.
  • Brackets are powder-coated with high-quality chemical-resistant epoxy powder
  • Constructed of 12-gauge steel
Shelf brackets provide a sturdy base of support for shelving units and are adjustable along vertical Atlas uprights.
Ordering information: Brackets are sold in sets of two. Low-profile shelf brackets and Kableway shelf brackets are designed for under mount only. For Atlas uprights, see VWRWMUP24, VWRC24, and VWRSU24 series. For Atlas shelving, see VWRNL1236, VWR1L1236, and VWR2L1236 series.