VWR® Shelf Label Holders
Supplier: VWR International
80094-296 80094-298 80094-300 80094-302 80094-304 80094-306
80094-296PK 212.54 CAD
80094-296 80094-298 80094-300 80094-302 80094-304 80094-306
VWR® Shelf Label Holders
Label Holders
Label holders are made of semi-rigid clear butyrate plastic that has been finished to eliminate glare for optimal barcode readability.
  • Semi-rigid plastic
  • Easy to exchange
  • Magnetic or self-adhesive
  • Finish eliminates glare
Label inserts are easily exchanged without removing the holder from its location. Self-adhesive holders will adhere permanently to any surface. Magnetic holders are easily repositioned on metal surfaces.
Ordering information: Label holders are provided with inserts that are compatible with laser and ink jet printers.