Synthware Gas Dispersion Tubes with 15° Angle, Fritted
Supplier: Kemtech America
F271018M F272025C F272025M F273035X F274045M
60003-616EA 90.95 CAD
60003-616 60003-620 60003-622 60003-624 60003-634
Synthware Gas Dispersion Tubes with 15° Angle, Fritted
Gas Tubes
The 15° angle allows tubes to be inserted into vessels when a large opening is not available.
  • Heavy wall borosilicate construction
  • Coarse, medium and extra coaurse frit porosity available
  • 10 mm disc
Tube stem ext. Ø is 8 mm. Stem length is 150 mm.