Gas Detection Pump Kit and Hydrogen Sulfide Detection Tubes, Sensidyne®
Supplier: Sensidyne
120SB 120SD 120SM 120U 830-0001-01
13502-596PK 659.56 CAD
13502-596 13502-610 13502-638 13502-650 14231-018 14236-032
Gas Detection Pump Kit and Hydrogen Sulfide Detection Tubes, Sensidyne®
Gas Detection Systems
This portable unit is lightweight, rugged, and capable of on-the-spot, precise detection of numerous gases and vapors
  • Measures gas and vapor concentrations in work area as easily as reading a thermometer
  • Calibration marks indicate gas concentration
  • Synthetic rubber sheath allows pump to be held firmly, even with wet gloves
  • Fail-safe handle locks in two positions (50 or 100cc)
  • Flow finish indicator makes viewing instantaneous and easy
Also available are hydrogen sulfide detection tubes with various measuring ranges for quick, accurate, and quantitativeMore Product Information