Genomic DNA Tissue Cell Kits, IBI Scientific
Supplier: IBI Scientific
Kits are designed specifically for purifying total DNA (including genomic, mitochondrial, and viral DNA) from a variety of animal tissue, paraffin-embedded tissue, buccal swab, and amniotic fluid
Note: For research use only.
The provided micropestle can efficiently homogenize tissue samples to shorten the time in the lysis step. Proteinase K and chaotropic salt are used to lyse cells and degrade protein, allowing DNA to be easily bound by the glass fiber matrix of the spin column. Once any contaminants have been removed, using a wash buffer (containing ethanol), the purified DNA is eluted by a low salt Elution Buffer or TE. The entire procedure can be completed within 1 hour without phenol/chloroform extraction or alcohol precipitation. The expected yield of genomic DNA is up to 50µg and the purified DNA (with approximately 20–30Kb) is suitable for use in PCR or other enzymatic reactions.
Caution: Components contain irritant agents. Always wear a lab coat, disposable gloves, and protective goggles during use.