Pedestal Autopsy Tables, Mortech®
Supplier: Avantor
29000-018EA 87698.3 CAD
29000-018 29000-020
Pedestal Autopsy Tables, Mortech®
Furniture Tables and Workbenches
All stainless steel tables feature heliarc-welded seams and joints with large inside and outside radii for easy and effective cleaning.
  • A removable panel allows for access to plumbing and electrical lines
  • Tables feature heavy-duty commercial grade ½ hp disposals and one waterproof GFCI electrical outlet
The elevating table features a hydraulic mechanical actuator and a heavy-duty internal scissor lift mechanism that raises or lowers the table by 15.2 cm (6").
Single-compartment sink basin measures 35.6W×30.5D×20.3H cm (14×12×8") and features a mixing faucet with a gooseneck spout and wrist blade handles. Tables are equipped with a hydro-aspirator with a cold water control valve and a vacuum breaker, as well as a spray hose assembly with a cold water control valve, nozzle, 3 m (10') of flexible hose, and a vacuum breaker. Plumbing and electrical lines are factory-installed and require only single-point connections at project locations.
Ordering information: Supplied with three 20.3 cm (8") wide solid or perforated stainless steel body supports. For optional features such as eyewash assemblies, gas/air valves, and post mortem scales, contact your Avantor representative.