accuro® Gas Detector Pump, Drager
Supplier: Dräger
accuro®
4056443 8317186 6400000 6400000 4056443
CA34330-072EA 3760.79 CAD
34330-072 CA34330-072 34330-070 CA34330-068 34330-068
accuro® Gas Detector Pump, Drager
Gas Detection Systems
For use with Short-term detector tubes (see 13453-238 series).
  • For use with Short-term detector tubes (see 13453-238 series)
  • Low-cost, one-handed bellows pump is ideal for measurements in hard-to-reach areas
  • Takes short-term measurements with reasonably few strokes when used with Draeger short-term detector tubes
This low-cost, one-handed bellows pump is ideal for measurements in hard-to-reach areas. It takes short-term measurements with reasonably few strokes when used with Draeger short-term detector tubes.
Ordering information: The accuro® gas detector pump is supplied with deluxe tube opener and Allen wrench. The accuro® hard side kit includes an accuro pump, deluxe tube opener, spare parts kit, 10 rubber caps, screwdriver, Allen wrench, and carrying case. The accuro® soft side kit includes an accuro® pump, deluxe tube opener, spare parts kit, screwdriver, Allen wrench, and nylon pouch.