E.Z.N.A.® Genomic DNA Isolation Kits, Omega Bio-Tek
Supplier: Omega Bio-Tek
E.Z.N.A®
D0926-00 D3591-00 D0926-01 D3350-02 D4015-02 D4015-01 D4015-01 D4015-02 D3350-02 D4015-00 D0926-02 D3591-02 D4015-00 D2492-03 D0926-02 D3591-01 D3350-00 D3350-00 D0926-00 D0926-01 D3350-01 D2492-03 D3591-02 D3350-01 D3591-00
CA101318-966EA 1181.49 CAD
CA101318-966 CA101318-922 CA101318-926 CA101318-924 CA101319-054 CA101319-056 101318-994 101318-992 101318-996 CA101318-882 101319-054 101319-056 101319-048 CA101319-344 CA101319-048 101318-882 101318-924 101318-922 101318-966 CA101318-992 101318-926 CA101318-996 101319-420 CA101318-994 101319-344
E.Z.N.A.® Genomic DNA Isolation Kits, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Genomic DNA kits allow for the isolation of genomic DNA from a variety of sample types including blood (Whole Blood and Related Body Fluids, Large-Scale), soil, stool, bacteria (Gram-Positive and Gram-Negative Bacterial Cultures ), yeast, insect (and invertebrate tissues), cultured cells, mouse tail, fungal and forensic samples (such as Dried Blood, Body Fluids, and Buccal Swabs).
Most kits are available in mini, midi, maxi, and 96-well formats. Isolated DNA can be used for restriction digest, PCR, southern blotting, and sequencing. Mag-Bind™ Genomic DNA kits can be adapted to most robotic liquid handling platforms.